Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Chicken Thyroxine ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of chicken T4.

$602.00

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 20 to 320ng/mL
Sensitivity 20ng/mL
Sample Type Plasma, Serum
Research Discipline Signal Transduction
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601511KT View Documents Novus Biologicals™
NBP2601511KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Chicken Thyroxine ELISA Kit (Colorimetric) measures chicken Thyroxine in serum, plasma.
Specifications

Specifications

ELISA Kit (Colorimetric)
20ng/mL
Signal Transduction
50 to 100 μL
T4
Thyroxine
ELISA
20 to 320ng/mL
Plasma, Serum
RUO
For detection of chicken T4
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Instruction manual, Standard, Stop Solution, Wash Buffer (20 x concentrate), Substrate B, Substrate A, HRP-avidin, Conjugate, Assay plate

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit