Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Chicken Triiodothyronine ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of chicken T3.

$602.00

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 0.5 to 8ng/mL
Sensitivity 0.5ng/mL
Sample Type Plasma, Serum, Tissue Homogenates
Research Discipline Signal Transduction
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601801KT View Documents Novus Biologicals™
NBP2601801KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Chicken Triiodothyronine ELISA Kit (Colorimetric) measures chicken Triiodothyronine in serum, plasma, tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
0.5ng/mL
Signal Transduction
50 to 100 μL
liothyronine; T3
Triiodothyronine
ELISA
0.5 to 8ng/mL
Plasma, Serum, Tissue Homogenates
RUO
For detection of chicken T3
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Adhesive Strip (For 96 wells), Assay plate, Conjugate, HRP-avidin, Wash Buffer (20 x concentrate), Substrate B, Substrate A, Standard, Stop Solution

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit