Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya Virus Mouse anti-Virus, Alexa Fluor™ 488, Clone: 6A11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP253111AF488

Catalog No. NP23111A488


Add to cart

Description

Description

Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, Immunofluorescence.
Specifications

Specifications

Chikungunya Virus
Monoclonal
Alexa Fluor 488
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
Virus
IgG2b
Western Blot, Immunofluorescence
6A11
Western Blot, Immunofluorescence
Chikungunya
Live chikungunya virus
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit