Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs

Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs are suitable for use with a variety of cooling vests.

$278.00

Specifications

Color Clear
Cooling Time Packs Maintain Constant 64°F (18°C) for up to 4 hours
For Use With (Application) Cooling Vest
Material Cotton with Gel Polymer
Product Type Cooling Pack
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-819-671
View Documents
Ergodyne™
12201
Small/Medium
Each for $278.00
Add to cart
 
19-819-672
View Documents
Ergodyne™
12202
Large/X-Large
Each for $278.00
Add to cart
 
Description

Description

  • Immediate and long-lasting cooling relief
  • Packs maintain constant 64°F (18°C) for up to 4 hours
  • Quick recharge in a little as 10 minutes
  • Patented honeycomb ice pack design eliminates stiffness
  • Wash by hand with a mild detergent and warm water
Specifications

Specifications

Clear
Cooling Vest
Cooling Pack
Packs Maintain Constant 64°F (18°C) for up to 4 hours
Cotton with Gel Polymer
6/Pk.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit