Learn More
Sonoco ThermoSafe Brands ChillTech™ Compact
Pre-qualified 5 day reusable shipping system for 2°C to 8°C. ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads.
Supplier:  Sonoco ThermoSafe Brands CHCM5U
 View more versions of this product
Catalog No. 03-600-321
Add to Cart
Description
Description
  • Pre-qualified range of 2°C to 8°C
  • Reusable and universal shipping systems
  • ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads
  • Simply change the outer insulated shipper to switch shipment duration from 48 hours to 72 hours to 144 hours
  • All solutions are fully qualified to ISTA 7D and 7E temperature profiles.
  • All components are replaceable
Specifications
Specifications
1 EACH
13.78 x 13.43 x9.76
8.78in. x 8.78in. x 4.25in.
5.4L
Shipper, gel packs, pads, payload box, and packout instructions
5-Day
COMPACT
VIP