Learn More
Sonoco ThermoSafe Brands ChillTech™ Global
Pre-qualified 6-day reusable shipping system for 2°C to 8°C. ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads.
Supplier:  Sonoco ThermoSafe Brands CHG10U
 View more versions of this product
Catalog No. 03-600-326
Add to Cart
Description
Description
  • Pre-qualified range of 2°C to 8°C
  • Reusable and universal shipping systems
  • ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads
  • Simply change the outer insulated shipper to switch shipment duration from 48 hours to 72 hours to 144 hours
  • All solutions are fully qualified to ISTA 7D and 7E temperature profiles.
  • All components are replaceable
Specifications
Specifications
1 EACH
15.657 x 15.59 x 15.67
8.78in. x 8.78in. x 9.19in.
10.3L
Shipper, gel packs, pads, payload box, and packout instructions
6-Day
GLOBAL
VIP AND EPS