Learn More
Sonoco ThermoSafe Brands ChillTech™ Global with ZERO Bench-Time™ technology
Pre-qualified 6-day reusable shipping system for 2°C to 8°C, with ZERO Bench-Time™ technology. ZERO Bench-Time™ technology allows for immediate packouts.  ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads.
Supplier:  Sonoco ThermoSafe Brands CHG16ZU
 View more versions of this product
Catalog No. 03-600-315
Add to Cart
Description
Description
  • Pre-qualified range of 2°C to 8°C
  • Reusable and universal shipping systems
  • ChillTech shippers utilize the same pack-out configuration across seasons, durations and payloads
  • Simply change the outer insulated shipper to switch shipment duration from 48 hours to 72 hours to 144 hours
  • Available with our patented ZERO Bench-Time® technology option which allows our solutions to be packed out immediately after the PCM bottles are removed from the freezer.
  • All solutions are fully qualified to ISTA 7D and 7E temperature profiles.
  • All components are replaceable
Specifications
Specifications
1 EACH
23.58 x 15.59 x 15.67
15.98in. x 8.11in. x 7.32in.
15.6L
Shipper, gel packs, pads, payload box, ZBT PACK and packout instructions
6-Day
GLOBAL + ZBT
VIP AND EPS