Learn More
Cayman Chemical CHIR99021, 252917-06-9, 1 mg
SDP
Supplier:  Cayman Chemical 131221
Encompass_Preferred
Molecular Formula C22H18Cl2N8, Purity ≥95%, Formula Weight 465.3, Synonyms: 6-[[2-[[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile, CT 99021; An aminopyrimidine derivative that inhibits GSK3 and GSK3 (IC50s = 10 and 6.7 nM, respectively); activates glycogen synthesis in CHO-IR cells (EC50 = 0.8 M) and in isolated type 1 diabetic rat skeletal muscle; has been shown to promote self-renewal of embryonic stem cells.
Catalog No. NC0699165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.