Learn More
Chiral Technologies, Inc. CHIRALPAK AD-3 4.6X 100
Supplier:  Chiral Technologies, Inc. 19523
Encompass
CHIRALPAK AD-3, 3 micron, 4.6 mm ID x 100 mm LN HPLC analytical column
Catalog No. 50-713-286