Learn More
Chiral Technologies, Inc. CHIRALPAK AGP 4X10 PKG/2  DFS Item
Manufacturer:  Chiral Technologies, Inc. 30711
Immobilized Protein-Based Columns, CHIRALPAK AGP, 5 micron, 4.0mm ID x 10mm LN, Guard Cartridges Pkg/2
Catalog No. 50-713-306