Learn More
Chiral Technologies, Inc. CHIRALPAK AS-3 4.6X 100  DFS Item
Manufacturer:  Chiral Technologies, Inc. 20523
CHIRALPAK AS-3, 3 micron, 4.6 mm ID x 100 mm LN HPLC analytical column
Catalog No. 50-713-290