Learn More
Chiral Technologies, Inc. CHIRALPAK AS-RH 2.1X150  DFS Item
Manufacturer:  Chiral Technologies, Inc. 20794
CHIRALPAK AS-RH, 5 micron, 2.1 mm ID x 150 mm LN HPLC analytical column
Catalog No. 50-713-363