Learn More
Chiral Technologies, Inc. CHIRALPAK IB-3 4X10 3PK  DFS Item
Manufacturer:  Chiral Technologies, Inc. 81511
Coated Polysaccharide,CHIRALPAK IB-3, 3 micron, 4.0 mm ID x 10 mm LN Guard Cartridges, 3PK
Catalog No. 50-713-272