Learn More
Chiral Technologies, Inc. CHIRALPAK IB 5UM 4X10 3PK  DFS Item
Manufacturer:  Chiral Technologies, Inc. 81311
Coated Polysaccharide, CHIRALPAK IB, 5 micron, 4.0 mm ID x 10 mm LN Guard Cartridges, 3PK
Catalog No. 50-713-267