Learn More
Chitinase 3-like 1 Rat anti-Human, Alexa Fluor™ 594, Clone: 9X24, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP221969AF594
Catalog No. NP221969A94
Add to cart
Description
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for ELISA.
Specifications
Specifications
Chitinase 3-like 1
9X24
ELISA
AW208766, Brp39, Chi3l1, gp39
Rat
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CHI3L1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Rat immunized with human recombinant protein of Chitinase 3-like 1. Rat anti human Chitinase 3-like 1 NP_001267.
0.1 mL
Primary
1116