Learn More
Chitinase 3-like 1 Rat anti-Human, Alexa Fluor™ 405, Clone: 9X24, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221969AF405
Catalog No. NP221969A40
Add to cart
Description
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for ELISA.
Specifications
Specifications
Chitinase 3-like 1
9X24
ELISA
AW208766, Brp39, Chi3l1, gp39
Rat
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
CHI3L1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Rat immunized with human recombinant protein of Chitinase 3-like 1. Rat anti human Chitinase 3-like 1 NP_001267.
0.1 mL
Primary
1116