Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitosan, molecular weight: 100,000-300,000, ACROS Organics™

Click to view available options
Quantity:
50g
100g
500g
Packaging:
Plastic bottle
9012-76-4
C56H103N9O39
1526.464
MFCD00161512
FLASNYPZGWUPSU-SICDJOISSA-N
chitosan,poliglusam,deacetylchitin,chicol,flonac c,flonac n,sea cure plus,kytex h,kytex m,kimitsu chitosan f
71853
methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O
9012-76-4
C56H103N9O39
1526.464
MFCD00161512
FLASNYPZGWUPSU-SICDJOISSA-N
chitosan,poliglusam,deacetylchitin,chicol,flonac c,flonac n,sea cure plus,kytex h,kytex m,kimitsu chitosan f
71853
methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O
Ash 2% max.
Physical Form Powder or Granules
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% max. (K.F.)
Color Beige-Yellow to White
Quantity 50g
Chemical Name or Material Chitosan, Molecular Weight: 100, 000 to 300, 000

Recommended Storage : Refrigerator +4°C