Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitosan, molecular weight: 100,000-300,000, ACROS Organics™
Click to view available options
Quantity:
50g
100g
500g
Packaging:
Plastic bottle
Specifications
Ash | 2% max. |
Physical Form | Powder or Granules |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Water | 10% max. (K.F.) |
Color | Beige-Yellow to White |
Quantity | 500g |
Chemical Name or Material | Chitosan, Molecular Weight: 100, 000 to 300, 000 |
Safety and Handling
Recommended Storage : Refrigerator +4°C