Learn More
Enzo Life SciencesSupplier Diversity Partner Chk 1 monoclonal antibody (DCS-310) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMCC111E
Clone: DCS-310. Host: Mouse. Isotype: IgG3. Immunogen: Recombinant human Chk1. UniProt: O14757. Species Reactivity: Human, Mouse, Rat. Applications: IHC, IP, WB. Recommended dilutions: Immunohistochemistry (1µg/ml). Immunoprecipitation (2µg/200µl cell extract, 5x106 cells). Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-444