Learn More
Enzo Life SciencesSupplier Diversity Partner Chk 2 monoclonal antibody (DCS-270) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMCC112E
Clone: DCS-270. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Chk2. UniProt: O96017. Species Reactivity: Human. Applications: IHC, IP, WB. Recommended dilutions: Immunohistochemistry (1-5µg/ml). Immunoprecipitation (3µg/200-300µl). Western Blot (0.2µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-445