Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Mouse anti-Human, Canine, Allophycocyanin/Cy7, Clone: OTI5C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271688APCCY7

Catalog No. N27168APCY7


Add to cart

Description

Description

Chk2 Monoclonal antibody specifically detects Chk2 in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Chk2
OTI5C4
Flow Cytometry
Affinity Purified, Conjugated
CHEK2
Full length recombinant protein of human Chk2 (NP_009125) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers
Primary
11200.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit