Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human CHL-1/L1CAM-2 ELISA Kit (Colorimetric)
SDP

Catalog No. NB230038
Click to view available options
Quantity:
1 Kit

Specifications

Assay Range 46.88-3000 pg/mL
Assay Sensitivity 16.9 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Human CHL-1/L1CAM-2.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 10752
Gene Symbol CHL1
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.