Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia LPS MAb Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABX84931KC
Description
Chlamydia LPS MAb Monoclonal antibody specifically detects Chlamydia LPS MAb in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Chlamydia LPS MAb | |
Monoclonal | |
Unconjugated | |
Mouse | |
Affinity Purified | |
Infectious Diseases | |
Specific for Chlamydia species LPS Reactive, A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, L3 and Chlamydia psittaci. | |
2 to 8°C. For long-term storage, freeze at -20°C. |
ELISA, Immunodiffusion, Western Blot | |
Clone 1 | |
Anti-Chlamydia LPS | |
Chlamydia Trachomatis LPS | |
1 mg | |
Primary | |
All species | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction