Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Lps Mouse anti-Bacteria, Clone: Tu-300, Novus Biologicals™

Mouse Monoclonal Antibody
$305.00
Specifications
Antigen | Chlamydia Lps |
---|---|
Clone | Tu-300 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062450
![]() |
Novus Biologicals
NB10062450 |
0.1 mg |
Each of 1 for $305.00
|
|
|||||
Description
Chlamydia Lps Monoclonal specifically detects Chlamydia Lps in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Chlamydia Lps | |
Monoclonal | |
Mouse | |
Bacteria | |
IgG2b | |
Affinity Purified | |
This product is specific for Chlamydia lipopolysaccharide (LPS). LPS is a common feature of the outer envelope of gram negative bacteria, which acts as a potent endotoxin, triggering an innate immune response. Studies have shown however, that whilst the LPS of Chlamydia trachomatis does evoke an immune response, it displays only weak endotoxic activity when compared to that of other bacteria such as Salmonella minnesota or Neisseria gonorrhoeae. This antibody has not been cross-adsorbed and may react with LPS from related species. |
Tu-300 | |
Unconjugated | |
RUO | |
Made to Bacterial CHLAMYDIA LPS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title