Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/284), Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $409.50
Specifications
Antigen | Chlamydia Trachomatis Lps |
---|---|
Clone | 1312/284 |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Description
Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Chlamydia Trachomatis Lps | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Chlamydia trachomatis L2 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1312/284 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Bacteria | |
IgG1 | |
Protein A purified | |
CHLAMYDIA TRACHOMATIS LPS |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title