Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorambucil, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
50 g
100 g
Packaging:
Amber Glass Bottle
305-03-3
C14H19Cl2NO2
304.21
JCKYGMPEJWAADB-UHFFFAOYSA-N
4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid
OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl
305-03-3
C14H19Cl2NO2
304.21
JCKYGMPEJWAADB-UHFFFAOYSA-N
4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid
OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl

C1659, 305-03-3, C14H19Cl2NO2

Spectrum™ Chlorambucil, USP is a chemotherapeutic drug that is currently used mainly in chronic lymphocytic leukemia. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 250 mg
Melting Range 65-69°C
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 101%