Learn More
Chloramine-T trihydrate, 98%, Thermo Scientific Chemicals
Description
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Notes
Incompatible with strong oxidizing agents, acids and ammonia.
Specifications
Melting Point | 167°C to 170°C |
pH | 8 to 10 |
Quantity | 5000 g |
Assay Percent Range | 98% |
UN Number | UN3263 |
Beilstein | 3924168 |
Flash Point | 192°C (377°F) |
Merck Index | 14,2075 |
Solubility Information | Soluble in water and ethyl alcohol. Insoluble in benzene and ethers. |
Formula Weight | 281.69 (227.67 Anhydrous) |
Show More |
Safety and Handling
Hazard Category | H302-H314-H334-H335 |
Hazard Statement | GHS H Statement H334-H314-H318-H302 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S. |
EINECSNumber | 204-854-7 |
RTECSNumber | XT5616800 |
TSCA | Yes |
storageNote1 | Moisture Sensitive |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.