Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine-T Trihydrate, Reagent, ACS, 98-103%, Spectrum™

Click to view available options
Quantity:
125 g
250 g
500 g
2.5 kg
12 kg
125 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7080-50-4
C7H7ClNO2S
204.65
VQWVWLDBJISZHX-UHFFFAOYSA-M
(chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate
CC1=CC=C(C=C1)S([O-])(=O)=NCl
7080-50-4
C7H7ClNO2S
204.65
VQWVWLDBJISZHX-UHFFFAOYSA-M
(chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate
CC1=CC=C(C=C1)S([O-])(=O)=NCl

CH120, 7080-50-4, C7H7SO2NClNa•3H2O

Spectrum™ Chloramine-T Trihydrate, Reagent, ACS is also known as tosylchloramide. It is most commonly used as a disinfectant or as a biocide. It is a white powder and can be a source of electrophilic chlorine in organic synthesis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Quantity 12 kg
Packaging Poly Pail
CAS Max % 100%
UN Number UN3263
Percent Purity 98-103%