Learn More
Chloramine-T Trihydrate, Reagent, ACS, 98-103%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
CH120, 7080-50-4, C7H7SO2NClNa•3H2O
Spectrum™ Chloramine-T Trihydrate, Reagent, ACS is also known as tosylchloramide. It is most commonly used as a disinfectant or as a biocide. It is a white powder and can be a source of electrophilic chlorine in organic synthesis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Packaging Fiber Drum