Learn More
Alkali Scientific Chloramphenicol, 1 Kg
SDP
Supplier:  Alkali Scientific CHL002CP
Encompass
Chloramphenicol is a broad-spectrum antibiotic provided in a 1 kilogram bulk laboratory format for large-scale microbiological and biochemical applications. It inhibits bacterial protein synthesis by binding to the 50S ribosomal subunit, effectively blocking peptide bond formation and bacterial growth. This compound is active against a wide range of Gram-positive and Gram-negative bacteria, making it suitable for use in selective media preparation and antimicrobial studies. The bulk packaging supports high-volume laboratory use and repeated experimental applications. Chloramphenicol is widely used in research laboratories for molecular biology, microbial selection, and culture media development requiring consistent antibacterial performance.
Catalog No. 50-006-60268