Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chloramphenicol, 10mg/mL in ethanol, sterile-filtered

Click to view available options
Quantity:
10mL
25mL
56-75-7
C11H12Cl2N2O5
323.126
MFCD00078159
WIIZWVCIJKGZOK-RKDXNWHRSA-N
5959
CHEBI:17698
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
56-75-7
C11H12Cl2N2O5
323.126
MFCD00078159
WIIZWVCIJKGZOK-RKDXNWHRSA-N
5959
CHEBI:17698
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
UN Number UN1987
Physical Form Liquid
Quantity 25mL
Chemical Name or Material Chloramphenicol, 10mg/mL in ethanol, sterile-filtered

GHS H Statement
H225-H350-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : AB6825000

TSCA : TSCA

storageNote1 : -30° to -10°C