Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine diacetate hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
206986-79-0
2C2H4O2·xH2O
625.55
MFCD00150042
JGFZDLOXFRFUTG-UHFFFAOYSA-N
chlorhexidine acetate hydrate 1:2:x,chlorhexidine diacetate salt monohydrate,chlorhexidine diacetate monohydrate,1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt,4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
43834499
acetic acid;(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;hydrate
CC(=O)O.CC(=O)O.C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.O
206986-79-0
2C2H4O2·xH2O
625.55
MFCD00150042
JGFZDLOXFRFUTG-UHFFFAOYSA-N
chlorhexidine acetate hydrate 1:2:x,chlorhexidine diacetate salt monohydrate,chlorhexidine diacetate monohydrate,1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt,4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
43834499
acetic acid;(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;hydrate
CC(=O)O.CC(=O)O.C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.O
Packaging Glass bottle
Loss on Drying 3.5% max. (105°C)
Color White
Melting Point 153.0°C to 157.0°C
Infrared Spectrum Authentic
Quantity 25g
Assay Percent Range 98%
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: soluble in alcohol, glycerol, propylene glycol and, polyethylene glycols
Formula Weight 625.55
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Chlorhexidine diacetate hydrate, 98%
Show More Show Less

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 200-302-4

RTECSNumber : DU1930000