Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloride Standard, 10 ppm Cl-, Ricca Chemical
Supplier: Ricca Chemical Company 1942.14
Specifications
CHLORIDE STANDARD | |
ClH4N | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
120 mL |
12125-02-9 | |
MFCD00011420 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
Natural Poly Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction