Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Standard, 5 ppm Cl, Reagents

Catalog No. 01335636
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Catalog No. Quantity
01-335-636 120 mL
01-335-635 1 L
01-335-634 500 mL
3 options

Catalog No. 01-335-636

Supplier: Reagents Holdings Llc CC155105120A

Only null left
Add to Cart
Add to Cart

Chloride Standard, 5 ppm Cl, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Chloride Standard, 5 ppm Cl
Packaging Poly Bottle
CAS 7732-18-5
Specific Gravity 1.0001
Physical Form Liquid
Molecular Formula NaCl/H2O
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 120 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.