Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorimuron ethyl, 95%

Click to view available options
Quantity:
1g
5g
25g
90982-32-4
C15H15ClN4O6S
414.817
MFCD00128063
NSWAMPCUPHPTTC-UHFFFAOYSA-N
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
56160
CHEBI:47319
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC
90982-32-4
C15H15ClN4O6S
414.817
MFCD00128063
NSWAMPCUPHPTTC-UHFFFAOYSA-N
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
56160
CHEBI:47319
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC
Merck Index 14,2093
Solubility Information Freely soluble in aqueous solution
Formula Weight 414.82
Physical Form Powder
Percent Purity 95%
Color Colorless
Melting Point 180°C to 182°C
Quantity 1g
Chemical Name or Material Chlorimuron ethyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG5095000

storageNote1 : 2° to 8°C