Learn More
Supply SolutionsSupplier Diversity Partner Hach - 2ml Chlorine Standard Solution; Platform: Solution; Volume: 2ml; Concentration: 50 - 75 mg/L Cl?; Description: Chlorine as CI2 (NIST); Quantity: pack of 20 PourRite Ampules DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 1426820
Chlorine standard solution is used for accuracy checks in Chlorine determinations. Lot-specific concentration displayed on label. Features: Simple Accuracy Checks: Voluette and PourRite ampule standards are designed to work directly with standard additions accuracy checks in procedures and photometer software, Minimize Waste: Open a fresh standard ampule for each accuracy check. Each ampule contains enough standard for one set of standard additions, Reliable Results: Sealed ampules prevent accidental contamination or evaporation of standard solutions.; Parameter: Multiple; Description: Chlorine as CI2 (NIST); Concentration: 50 mgL to 75 mgL Cl2; Volume: 20 pk - 2 mL PourRite Ampules; Platform : Solution; Minimize waste; Reliable results
Catalog No. 50-476-364
Due to product restrictions, please to purchase or view availability for this product.