Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorine Standard Solution, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Grade Chemical Solution
CAS 7782-50-5,7732-18-5,7758-05-6
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.