Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorine Water, Saturated Solution, Approx. 0.4% Chlorine, U.S.P. Chlorine Test Solution, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

C-205, 7782-50-5

Spectrum™ Chlorine Water, Saturated Solution, Approx. 0.4 Percent Chlorine, U.S.P. Chlorine Test Solution is a Spectrum solution concentrate that allows the preparation of fresh, reliable standard volumetric solutions quickly and accurately. Simply dilute the desired USP test solution concentrate to the specified volume (carefully following instructions provided with the package) and the resulting solution will have an exact Normality within 0.1%. Spectrum USP test solution concentrates are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications.
CAS 7782-50-5,7732-18-5
CAS Max % 0.004,0.996
Molecular Formula Cl2
UN Number UN1760
InChI Key KZBUYRJDOAKODT-UHFFFAOYSA-N
SMILES ClCl
IUPAC Name dichlorine
Molecular Weight (g/mol) 70.90
Packaging Amber Glass Bottle
Quantity 500 mL