Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlormezanone, 98+%

Click to view available options
Quantity:
25g
80-77-3
C11H12ClNO3S
273.731
MFCD00143951
WEQAYVWKMWHEJO-UHFFFAOYSA-N
chlormezanone,chlormethazanone,chlormethazone,trancopal,chlormezanon,clormetazanone,clormetazon,phenarol,dl-chlormezanone,clorilax
2717
CHEBI:3619
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
CN1C(S(=O)(=O)CCC1=O)C2=CC=C(C=C2)Cl
80-77-3
C11H12ClNO3S
273.731
MFCD00143951
WEQAYVWKMWHEJO-UHFFFAOYSA-N
chlormezanone,chlormethazanone,chlormethazone,trancopal,chlormezanon,clormetazanone,clormetazon,phenarol,dl-chlormezanone,clorilax
2717
CHEBI:3619
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
CN1C(S(=O)(=O)CCC1=O)C2=CC=C(C=C2)Cl

Skeletal muscle relaxant

Melting Point 114°C
Color White
Quantity 25g
Merck Index 14,2106
Formula Weight 273.74
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material Chlormezanone

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XJ1050000