Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlormezanone, Tocris Bioscience™

Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
80-77-3
C11H12ClNO3S
273.731
WEQAYVWKMWHEJO-UHFFFAOYSA-N
chlormezanone, chlormethazanone, chlormethazone, trancopal, chlormezanon, clormetazanone, clormetazon, phenarol, dl-chlormezanone, clorilax
2717
CHEBI:3619
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
CN1C(S(=O)(=O)CCC1=O)C2=CC=C(C=C2)Cl
This item is not returnable. View return policy
80-77-3
C11H12ClNO3S
273.731
WEQAYVWKMWHEJO-UHFFFAOYSA-N
chlormezanone, chlormethazanone, chlormethazone, trancopal, chlormezanon, clormetazanone, clormetazon, phenarol, dl-chlormezanone, clorilax
2717
CHEBI:3619
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
CN1C(S(=O)(=O)CCC1=O)C2=CC=C(C=C2)Cl

Skeletal muscle relaxant

Anxiolytic and skeletal muscle relaxant that acts at the benzodiazepine site of GABAA receptors.
Formula Weight 273.73
Percent Purity >99%
Quantity 1 g
Chemical Name or Material Chlormezanone

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit