Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro bis-(4-fluorophenyl)methane, 98%, Thermo Scientific™

Catalog Number AC134960100
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 88°C (0.1 Mbar)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5560 to 1.558
Quantity 10 g
Specific Gravity 1.25
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone
Formula Weight 238.66
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloro bis-(4-fluorophenyl)methane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-201-4

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit