Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro bis-(4-fluorophenyl)methane, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
Packaging:
Glass bottle
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride,4,4'-chloromethylene bis fluorobenzene,1,1'-chloromethylene bis 4-fluorobenzene,chlorobis 4-fluorophenyl methane,4,4'-difluorodiphenylmethylchloride,chloro bis-4-fluorophenyl methane,bis 4-fluorophenyl chloromethane,4,4'-difluorobenzhydrylchloride,1-chloro 4-fluorophenyl methyl-4-fluorobenzene,1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride,4,4'-chloromethylene bis fluorobenzene,1,1'-chloromethylene bis 4-fluorobenzene,chlorobis 4-fluorophenyl methane,4,4'-difluorodiphenylmethylchloride,chloro bis-4-fluorophenyl methane,bis 4-fluorophenyl chloromethane,4,4'-difluorobenzhydrylchloride,1-chloro 4-fluorophenyl methyl-4-fluorobenzene,1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
Density 1.2500g/mL
Specific Gravity 1.25
Flash Point 81°C
Boiling Point 88°C (0.1 mbar)
Refractive Index 1.5560 to 1.558
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone
Formula Weight 238.66
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Chloro bis-(4-fluorophenyl)methane, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 248-201-4