Alfa Aesar™ Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

Manufacturer: Alfa Aesar™ 4643003

Quantity 1g

 View more versions of this product

Catalog No. AA4643003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32993-05-8
    Formula Weight 772.27
    MDL Number MFCD00075004
    Synonym Cyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp(PPh3)
    Chemical Name or Material Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

    Specifications

    Formula Weight 772.27
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 135°C (dec.)
    Physical Form Crystalline Solid
    Synonym Cyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp(PPh3)
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00075004
    CAS 32993-05-8
    Color Orange
    Quantity 1g
    Chemical Name or Material Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

    GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit