Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chloro(cyclopentadienyl)(triphenylphosphine)nickel(II), Acros Organics

Click to view available options
Quantity:
1g
31904-79-7
C23H25ClNiP-
426.57
MFCD02093655
DYZCJABKDYGEDF-UHFFFAOYSA-M
chloronickel,cyclopentane,triphenylphosphane
134128029
cyclopentane;nickel;triphenylphosphane;chloride
C1CCCC1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ni]
31904-79-7
C23H25ClNiP-
426.57
MFCD02093655
DYZCJABKDYGEDF-UHFFFAOYSA-M
chloronickel,cyclopentane,triphenylphosphane
134128029
cyclopentane;nickel;triphenylphosphane;chloride
C1CCCC1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ni]
Quantity 1g
Color Bordeaux
Infrared Spectrum Authentic
Formula Weight 421.55
Physical Form Crystalline Powder
Chemical Name or Material chloro(cyclopentadienyl)(triphenylphosphine)nickel(II)