Alfa Aesar™ Chloro(dimethyl)phosphine, 97%

Manufacturer: Alfa Aesar™ 3016002

Quantity 0.5g

 View more versions of this product

Catalog No. AA3016002

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 811-62-1
    Linear Formula (CH3)2PCl
    Formula Weight 96.5
    MDL Number MFCD00013620
    UN Number UN2845
    Chemical Name or Material Chloro(dimethyl)phosphine

    Specifications

    Formula Weight 96.5
    Health Hazard 2 GHS H Statement
    H225-H250-H252-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −18° (−1°F)
    Boiling Point 76° to 77°C
    Quantity 0.5g
    Physical Form Liquid
    Linear Formula (CH3)2PCl
    MDL Number MFCD00013620
    UN Number UN2845
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Density 1.22g/mL
    Odor Pungent
    Sensitivity Air & Moisture Sensitive
    Color Colorless
    CAS 811-62-1
    Chemical Name or Material Chloro(dimethyl)phosphine
    Assay 97%

    GHS H Statement
    H225-H250-H252-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit