Alfa Aesar™ Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)

Manufacturer: Alfa Aesar™ 4662903

Quantity 1g

 View more versions of this product

Catalog No. AA4662903

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 92361-49-4
    Formula Weight 796.33
    MDL Number MFCD09038426
    Synonym Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp*(PPh{3})2
    Chemical Name or Material Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)

    Specifications

    Synonym Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp*(PPh{3})2
    Formula Weight 796.33
    MDL Number MFCD09038426
    Storage Note 1 2° to 8°C
    Sensitivity Air Sensitive
    Melting Point 128°C (dec.)
    Color Yellow
    Physical Form Crystalline Solid
    CAS 92361-49-4
    Quantity 1g
    Chemical Name or Material Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit