Alfa Aesar™ Chloroacetaldehyde, 45% w/w in water

Manufacturer: Alfa Aesar™ A11037-0I

Quantity 5000g

 View more versions of this product

Catalog No. AAA110370I

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 107-20-0
    Molecular Formula C2H3ClO
    Linear Formula ClCH2CHO
    Formula Weight 78.5
    MDL Number MFCD00006992
    UN Number UN2232
    Beilstein 1071226
    Merck Index 14,2109
    Synonym 2-Chloroethanal
    Chemical Name or Material Chloroacetaldehyde

    Specifications

    Formula Weight 78.5
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Odor Pungent
    UN Number UN2232
    Molecular Formula C2H3ClO
    Linear Formula ClCH2CHO
    Synonym 2-Chloroethanal
    Health Hazard 1 Danger
    RTECS Number AB2450000
    Flash Point 62°C (143°F)
    pH 1.2
    TSCA TSCA
    MDL Number MFCD00006992
    CAS 107-20-0
    Merck Index 14,2109
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H314-H318-H351-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Com
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: 2-CHLOROETHANAL
    Melting Point −28°C
    Beilstein 1071226
    Concentration 45% w/w in water
    Boiling Point 80° to 100°C
    Quantity 5000g
    Chemical Name or Material Chloroacetaldehyde

    GHS H Statement
    H301-H311-H330-H314-H318-H351-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Com

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit