Alfa Aesar™ Chloroacetaldehyde, 45% w/w in water

Manufacturer: Alfa Aesar™ A1103730

Quantity 250g

 View more versions of this product

Catalog No. AAA1103730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-20-0
    Molecular Formula C2H3ClO
    Linear Formula ClCH2CHO
    Formula Weight 78.5
    MDL Number MFCD00006992
    UN Number UN2232
    Beilstein 1071226
    Merck Index 14,2109
    Synonym 2-Chloroethanal
    Chemical Name or Material Chloroacetaldehyde

    Specifications

    Formula Weight 78.5
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H314-H318-H351-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    UN Number UN2232
    Quantity 250g
    CAS 107-20-0
    Synonym 2-Chloroethanal
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: 2-CHLOROETHANAL
    Concentration 45% w/w in water
    Flash Point 62°C (143°F)
    Boiling Point 80° to 100°C
    Melting Point −28°C
    TSCA TSCA
    Odor Pungent
    Health Hazard 1 Danger
    RTECS Number AB2450000
    pH 1.2
    Molecular Formula C2H3ClO
    Linear Formula ClCH2CHO
    MDL Number MFCD00006992
    Beilstein 1071226
    Merck Index 14,2109
    Chemical Name or Material Chloroacetaldehyde

    Description

    Applications
    Chloroacetaldehyde is a useful intermediate in the synthesis, it is also used to facilitate bark removal from tree trunks.

    Solubility
    soluble in acetone, methanol. Fully miscible in water.

    Notes
    Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

    GHS H Statement
    H301-H311-H330-H314-H318-H351-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Combustible liquid.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit