Alfa Aesar™ Chloroacetaldehyde dimethyl acetal, 98%

Manufacturer: Alfa Aesar™ B2504922

Quantity 100g

 View more versions of this product

Catalog No. AAB2504922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97-97-2
    Molecular Formula CICH2CH(OCH3)2
    Formula Weight 124.57
    MDL Number MFCD00000948
    UN Number UN1993
    Beilstein 1733700
    Synonym 2-Chloro-1,1-dimethoxyethane; Dimethyl chloroacetal
    Chemical Name or Material Chloroacetaldehyde dimethyl acetal

    Specifications

    Formula Weight 124.57
    Odor Aromatic
    UN Number UN1993
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H226-H332
    Toxic if swallowed.
    Flammable liquid and vapour.
    Harmful if inhaled.
    Flash Point 28°C (82°F)
    Boiling Point 128° to 130°C
    Melting Point <−73°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula CICH2CH(OCH3)2
    MDL Number MFCD00000948
    CAS 97-97-2
    Beilstein 1733700
    Synonym 2-Chloro-1,1-dimethoxyethane; Dimethyl chloroacetal
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Chloroacetaldehyde dimethyl acetal
    Assay 98%

    GHS H Statement
    H301-H226-H332
    Toxic if swallowed.
    Flammable liquid and vapour.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit