Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, 99+%, pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
108-90-7
C6H5Cl
112.556
MFCD00000530
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl
108-90-7
C6H5Cl
112.556
MFCD00000530
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl
Additional Information Vapor Pressure: 2mbar at 20°C
Assay Percent Range 99+%
Linear Formula C6H5Cl
Beilstein 05, 199
Merck Index 15, 2122
Formula Weight 112.56
Physical Form Liquid
Percent Purity ≥99%
Grade Pure
Density 1.1075g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.5230 to 1.5250
Solubility Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Specific Gravity 1.1075
Flash Point 23°C
Viscosity 0.8 mPa.s (20°C)
Boiling Point 131.0°C
Color Undesignated
Melting Point -45.0°C
Quantity 2.5L
Chemical Name or Material Chlorobenzene
Show More Show Less

GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.

GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning